Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026555.2 | NP_080831.2 | 5, 6 | 803 | ||
GenBank mRNA | AK003918.1 | - | 4, 5 | 473 | ||
AK077943.1 | - | 5, 6 | 743 | |||
AK082762.1 | - | 5, 6 | 733 | |||
AK086705.1 | - | 5, 6 | 743 | |||
AK132143.1 | - | 5, 6 | 737 | |||
BC005487.1 | - | 4, 5 | 640 | |||
BC025602.1 | - | 5, 6 | 754 | 5250505 | ||
BC055903.1 | - | 5, 6 | 750 | 4010685 | ||
BC092069.1 | - | 5, 6 | 701 | 4215821 |