Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177469.3 | NP_803420.2 | 8 | 2335 | ||
GenBank mRNA | AB096262.1 | - | 6 | 1398 | ||
AK034188.1 | - | 7 | 1898 | |||
AK082207.1 | - | 8 | 2214 | |||
BC056493.1 | - | 6 | 1619 | 5704932 | ||
BC094311.1 | - | 3 | 883 |
Set Membership: |
G-protein coupled receptor |