Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026391.2 | NP_080667.1 | 5, 6 | 599 | ||
GenBank mRNA | AF366393.1 | - | 5, 6 | 507 | ||
AK005033.1 | - | 4, 5 | 488 | |||
AK011198.1 | - | 5, 6 | 621 | |||
AK129386.2 | - | 5, 6 | 626 | |||
BC066022.1 | - | 5, 6 | 599 | 6828707 |