Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198831.2 | NP_942128.2 | 5 | 472 | ||
GenBank mRNA | AK007653.1 | - | 5 | 743 | ||
AK171917.1 | - | 5 | 445 | |||
BC037190.1 | - | 5 | 427 | 4168034 | ||
BC096562.1 | - | 3 | 269 | |||
BC116358.1 | - | 4 | 342 | 40099962 | ||
BC116359.1 | - | 4 | 339 | 40099963 | ||
BC128487.1 | - | 4 | 342 | 40099964 | ||
BC128488.1 | - | 4 | 342 | 40099965 | ||
BG077486.2 | - | 4 | 306 | |||
BU604260.1 | - | 5 | 465 | |||
CD363582.1 | - | 5 | 403 | |||
DN174444.1 | - | 5 | 472 |