Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199020.1 | NP_001185949.1 | 3 | 331 | ||
NM_021421.4 | NP_067396.3 | 2 | 466 | |||
GenBank mRNA | AK017545.1 | - | 2 | 429 | ||
AK077039.1 | - | 2 | 412 | |||
AK135709.1 | - | 3 | 328 | |||
AK158731.1 | - | 2 | 482 | |||
AK166619.1 | - | 3 | 330 | |||
BC024907.1 | - | 2 | 426 | 2812124 |