Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002260.3 | NP_002251.2 | 1 | 61 | ||
GenBank mRNA | AF260134.1 | - | 1 | 54 | ||
AK295525.1 | - | 1 | 49 | |||
BC093644.1 | - | 1 | 85 | 7939489 | ||
BC106069.1 | - | 1 | 13 | |||
BC112039.1 | - | 1 | 97 | 8327507 | ||
X54869.1 | - | 1 | 61 | |||
Y13055.1 | - | 1 | 61 |
Gene Symbol: KLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF350016.1 | - | 1 | 54 | ||
AF461157.1 | - | 1 | 70 |