Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026538.3 | NP_080814.1 | 5, 6 | 660 | ||
GenBank mRNA | AK011136.1 | - | 5, 6 | 670 | ||
AK076047.1 | - | 5, 6 | 678 | |||
AK076975.1 | - | 5, 6 | 660 | |||
AK077514.1 | - | 5, 6 | 673 | |||
AK145628.1 | - | 5, 6 | 701 | |||
AK159793.1 | - | 5, 6 | 664 | |||
AK169654.1 | - | 5, 6 | 675 | |||
BC018291.1 | - | 5, 6 | 654 | 4190167 |