Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002260.3 | NP_002251.2 | 3 | 326 | ||
GenBank mRNA | AF260134.1 | - | 3 | 319 | ||
AK295525.1 | - | 3 | 314 | |||
BC093644.1 | - | 3 | 350 | 7939489 | ||
BC106069.1 | - | 3 | 278 | |||
BC112039.1 | - | 3 | 362 | 8327507 | ||
X54869.1 | - | 3 | 326 | |||
Y13055.1 | - | 3 | 326 |
Gene Symbol: KLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF078550.1 | - | 3 | 319 | ||
AF350016.1 | - | 3 | 319 | |||
AF350017.1 | - | 3 | 319 | |||
AF461157.1 | - | 3 | 335 | |||
AK307860.1 | - | 3 | 212 | |||
AK311162.1 | - | 3 | 241 | |||
L14542.1 | - | 3 | 364 |