Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 3 | 300 | ||
NM_005040.3 | NP_005031.1 | 4 | 662 | |||
NM_199418.3 | NP_955450.2 | 5 | 725 | |||
XM_005274093.1 | XP_005274150.1 | 4 | 392 | |||
GenBank mRNA | AB451270.1 | - | 4 | 461 | ||
AB451397.1 | - | 4 | 461 | |||
AK091786.1 | - | 5 | 552 | |||
AK302386.1 | - | 3 | 251 | |||
AK312919.1 | - | 4 | 483 | |||
BC001500.2 | - | 4 | 480 | 3510081 | ||
BI827978.1 | - | 4 | 654 | |||
DB028823.1 | - | 4 | 442 | |||
L13977.1 | - | 4 | 490 |