Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163018.1 | NP_001156490.1 | 5, 6 | 750 | ||
NM_199196.2 | NP_954666.1 | 6, 7 | 819 | |||
GenBank mRNA | AK122213.1 | - | 6, 7 | 804 | ||
BC064461.1 | - | 6, 7 | 819 | 6821922 | ||
BC084591.1 | - | 1, 2 | 90 |