Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145940.2 | NP_666052.1 | 5 | 579 | ||
GenBank mRNA | AK031672.1 | - | 5 | 588 | ||
AK034907.1 | - | 5 | 623 | |||
AK045131.1 | - | 5 | 589 | |||
AK050495.1 | - | 5 | 587 | |||
AK159608.1 | - | 5 | 608 | |||
AK170563.1 | - | 5 | 623 | |||
AK186984.1 | - | 3 | 251 | |||
BC024811.1 | - | 5 | 547 | |||
BC024883.1 | - | 5 | 543 | |||
BC025560.1 | - | 5 | 579 | 5322999 |