Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166413.1 | NP_001159885.1 | 8, 9 | 862 | ||
NM_028027.3 | NP_082303.2 | 8, 9 | 924 | |||
GenBank mRNA | AF487515.1 | - | 8, 9 | 890 | ||
AK010452.1 | - | 8, 9 | 890 | |||
AK158684.1 | - | 8, 9 | 862 |