Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115132.2 | NP_001108604.1 | 7 | 689 | ||
NM_001271600.1 | NP_001258529.1 | 7 | 596 | |||
NM_001271601.1 | NP_001258530.1 | 7 | 689 | |||
GenBank mRNA | AK002487.1 | - | 7 | 607 | ||
AK037175.1 | - | 7 | 640 | |||
AK080447.1 | - | 7 | 628 | |||
AK083981.1 | - | 7 | 547 | |||
AK085584.1 | - | 7 | 620 | |||
AK088294.1 | - | 7 | 629 | |||
AK146642.1 | - | 7 | 673 | |||
AK152158.1 | - | 7 | 657 | |||
AK153528.1 | - | 7 | 629 | |||
AK163958.1 | - | 7 | 619 | |||
AK165250.1 | - | 7 | 640 | |||
BC003900.1 | - | 7 | 628 | 3601298 |