Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115132.2 | NP_001108604.1 | 5 | 508 | ||
NM_001271600.1 | NP_001258529.1 | 5 | 415 | |||
NM_001271601.1 | NP_001258530.1 | 5 | 508 | |||
GenBank mRNA | AK002487.1 | - | 5 | 426 | ||
AK037175.1 | - | 5 | 459 | |||
AK080447.1 | - | 5 | 447 | |||
AK083981.1 | - | 5 | 366 | |||
AK085584.1 | - | 5 | 439 | |||
AK088294.1 | - | 5 | 448 | |||
AK146642.1 | - | 5 | 492 | |||
AK152158.1 | - | 5 | 476 | |||
AK153528.1 | - | 5 | 448 | |||
AK163958.1 | - | 5 | 438 | |||
AK165250.1 | - | 5 | 459 | |||
BC003900.1 | - | 5 | 447 | 3601298 |