Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026559.3 | NP_080835.1 | 3 | 318 | ||
GenBank mRNA | AJ344103.1 | - | 3 | 307 | ||
AK004219.1 | - | 3 | 326 | |||
AK007595.1 | - | 3 | 322 | |||
AK013103.1 | - | 3 | 261 | |||
AK076425.1 | - | 3 | 318 | |||
AK169882.1 | - | 3 | 261 | |||
BC030344.1 | - | 3 | 316 | 3673521 |