Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252582.1 | NP_001239511.1 | 3 | 658 | ||
NM_001252583.1 | NP_001239512.1 | 3 | 623 | |||
NM_144833.3 | NP_659082.1 | 3 | 623 | |||
GenBank mRNA | AK033090.1 | - | 3 | 400 | ||
AK042224.1 | - | 3 | 380 | |||
AK049384.1 | - | 4 | 678 | |||
AK053001.1 | - | 3 | 348 | |||
AK083003.1 | - | 3 | 365 | |||
AK132345.1 | - | 3 | 361 | |||
AK139797.1 | - | 2 | 177 | |||
BC021528.1 | - | 3 | 378 | 5353673 | ||
BC052080.1 | - | 3 | 355 | 6403411 | ||
CK791111.1 | - | 3 | 623 |