Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130107.1 | NP_001123579.1 | 12 | 1781 | ||
NM_005552.4 | NP_005543.2 | 12 | 1781 | |||
NM_182923.3 | NP_891553.2 | 12 | 1781 | |||
GenBank mRNA | AB451299.1 | - | 11 | 1461 | ||
AB451431.1 | - | 11 | 1461 | |||
AK294508.1 | - | 12 | 1766 | |||
AY180163.1 | - | 2 | 145 | |||
AY180164.2 | - | 12 | 1728 | |||
AY180165.3 | - | 2 | 145 | |||
AY180166.1 | - | 2 | 145 | |||
AY180167.1 | - | 2 | 145 | |||
AY180168.1 | - | 2 | 145 | |||
AY180169.2 | - | 2 | 145 | |||
AY180170.1 | - | 2 | 145 | |||
AY244715.5 | - | 2 | 145 | |||
BC008881.2 | - | 12 | 1742 | 4301579 | ||
L04733.1 | - | 12 | 1737 |