Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029836.3 | NP_084112.1 | 5 | 1317 | ||
GenBank mRNA | AK018700.1 | - | 5 | 1259 | ||
AY273809.1 | - | 5 | 1158 | |||
BC004006.1 | - | 5 | 965 | |||
BC030334.1 | - | 3 | 464 | |||
BC043694.1 | - | 4 | 1178 | |||
BC054393.1 | - | 5 | 1237 | 6491672 |