Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027570.3 | NP_081846.3 | 8 | 1032 | ||
GenBank mRNA | AK014649.1 | - | 8 | 1038 | ||
AK029820.1 | - | 1446 | ||||
AK037996.1 | - | 342 | ||||
AY092768.1 | - | 8 | 1037 | |||
BC039155.1 | - | 8 | 1028 | 4242682 | ||
BC055443.1 | - | 8 | 1032 | 4235345 |