Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbndd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048227.1 | NP_001041692.1 | 3 | 324 | ||
NM_001048228.1 | NP_001041693.1 | 3 | 348 | |||
NM_001048229.1 | NP_001041694.1 | 2 | 351 | |||
NM_026797.2 | NP_081073.1 | 3 | 453 | |||
GenBank mRNA | AK003735.1 | - | 3 | 349 | ||
AK013571.1 | - | 3 | 259 | |||
AY187289.1 | - | 3 | 332 | |||
BC019947.1 | - | 3 | 355 | 4022474 | ||
BC030298.1 | - | 3 | 602 | 5364949 | ||
BG080686.2 | - | 3 | 324 | |||
CJ156083.1 | - | 2 | 353 | |||
CX241653.1 | - | 3 | 453 |