Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083328.1 | NP_001076797.1 | 4 | 359 | ||
NM_001083329.1 | NP_001076798.1 | 3 | 265 | |||
NM_028422.1 | NP_082698.1 | 4 | 404 | |||
NM_181424.3 | NP_852089.2 | 5 | 498 | |||
GenBank mRNA | AK007444.1 | - | 2 | 353 | ||
AK040689.1 | - | 4 | 361 | |||
AK163448.1 | - | 5 | 509 | |||
AK201295.1 | - | 1 | 70 | |||
AW647214.1 | - | 2 | 229 | |||
BC025144.1 | - | 5 | 497 | 4483063 | ||
BC028518.1 | - | 2 | 243 | |||
BF012021.1 | - | 3 | 283 | |||
BY726952.1 | - | 5 | 509 | |||
CX210755.1 | - | 3 | 387 |