Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083328.1 | NP_001076797.1 | 4 | 398 | ||
NM_001083329.1 | NP_001076798.1 | 3 | 304 | |||
NM_028422.1 | NP_082698.1 | 4 | 443 | |||
NM_181424.3 | NP_852089.2 | 5 | 537 | |||
GenBank mRNA | AK007444.1 | - | 2 | 392 | ||
AK040689.1 | - | 4 | 400 | |||
AK163448.1 | - | 5 | 548 | |||
AK201295.1 | - | 1 | 109 | |||
AW647214.1 | - | 2 | 268 | |||
BC025144.1 | - | 5 | 536 | 4483063 | ||
BC028518.1 | - | 2 | 282 | |||
BF012021.1 | - | 3 | 322 | |||
BY726952.1 | - | 5 | 548 | |||
DT929445.1 | - | 1 | 21 |