Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdipt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026638.3 | NP_080914.1 | 6 | 1439 | ||
GenBank mRNA | AK076974.1 | - | 6 | 1439 | ||
AK079226.1 | - | 6 | 916 | |||
AK089109.1 | - | 6 | 1005 | |||
AK132286.1 | - | 6 | 1025 | |||
AK215144.1 | - | 1 | 73 | |||
BC021473.1 | - | 6 | 967 | 5343411 | ||
BC024413.1 | - | 6 | 779 | 4973307 | ||
BC032182.1 | - | 6 | 976 | 5252470 |