Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177776.1 | NP_001171247.1 | 3 | 438 | ||
NM_026203.3 | NP_080479.2 | 20 | 3221 | |||
GenBank mRNA | AK036336.1 | - | 20 | 3111 | ||
AK161335.1 | - | 3 | 439 | |||
AY133241.1 | - | 20 | 3114 | |||
AY133242.1 | - | 5 | 499 | |||
BC038502.1 | - | 13 | 1572 | |||
BC055400.1 | - | 19 | 2943 | 6467369 | ||
BC065146.1 | - | 20 | 3116 |