Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016749.2 | NP_058029.2 | 6, 7 | 1013 | ||
GenBank mRNA | BC116684.1 | - | 6, 7 | 980 | 40106746 | |
BC118050.1 | - | 6, 7 | 980 | 40106744 | ||
CB587371.1 | - | 2, 3 | 260 | |||
U68267.1 | - | 6, 7 | 1007 |