Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF534114.1 | - | 2 | 155 | ||
AK018166.1 | - | 3 | 206 | |||
AK086301.1 | - | 3 | 365 | |||
AK134581.1 | - | 3 | 209 | |||
AK153400.1 | - | 3 | 193 | |||
AK208385.1 | - | 1 | 13 | |||
BC005640.1 | - | 3 | 194 | |||
BC018178.1 | - | 3 | 180 | |||
BC037451.1 | - | 2 | 184 | 4018648 | ||
U27462.1 | - | 3 | 402 |