Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016771.3 | NP_058051.3 | 3 | 443 | ||
GenBank mRNA | AF026073.1 | - | 3 | 321 | ||
AI118913.1 | - | 3 | 455 | |||
AK133530.1 | - | 3 | 362 | |||
AK160466.1 | - | 3 | 428 | |||
BC010752.1 | - | 3 | 416 | |||
BC066190.1 | - | 3 | 396 | 30472198 | ||
U32371.1 | - | 3 | 393 |