Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256269.1 | NP_001243198.1 | 7 | 966 | ||
NM_001256270.1 | NP_001243199.1 | 6 | 1054 | |||
NM_007317.2 | NP_015556.1 | 6 | 917 | |||
GenBank mRNA | AB017430.2 | - | 6 | 894 | ||
AK223431.1 | - | 6 | 903 | |||
AK294380.1 | - | 6 | 1293 | |||
AK297893.1 | - | 5 | 590 | |||
AK312234.1 | - | 6 | 908 | |||
AK316050.1 | - | 6 | 1054 | |||
AK316389.1 | - | 7 | 925 | |||
BC004352.1 | - | 6 | 889 | 3535435 | ||
BC028155.1 | - | 6 | 875 | 5241557 | ||
BT007259.1 | - | 6 | 870 |