Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109748.1 | NP_001103218.1 | 2 | 482 | ||
NM_016871.2 | NP_058567.2 | 3 | 384 | |||
GenBank mRNA | AF043249.1 | - | 2 | 310 | ||
AF316403.1 | - | 2 | 317 | |||
AF316404.1 | - | 2 | 317 | |||
AK142575.1 | - | 2 | 78 | |||
AK150019.1 | - | 2 | 479 | |||
AK163759.1 | - | 3 | 368 | |||
AK164222.1 | - | 3 | 383 | |||
AK164570.1 | - | 3 | 368 | |||
AK165173.1 | - | 3 | 379 | |||
AK170991.1 | - | 3 | 368 | |||
AK179196.1 | - | 46 | ||||
AK182800.1 | - | 2 | 63 | |||
AK187273.1 | - | 1 | 46 | |||
AK215349.1 | - | 1 | 46 | |||
BC100452.1 | - | 3 | 372 | 6532792 |