Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256269.1 | NP_001243198.1 | 10, 11 | 1556 | ||
NM_001256270.1 | NP_001243199.1 | 9, 10 | 1644 | |||
NM_007317.2 | NP_015556.1 | 9, 10 | 1507 | |||
GenBank mRNA | AB017430.2 | - | 9, 10 | 1484 | ||
AK223431.1 | - | 9, 10 | 1493 | |||
AK294380.1 | - | 9, 10 | 1883 | |||
AK312234.1 | - | 9, 10 | 1498 | |||
AK316050.1 | - | 9, 10 | 1644 | |||
AK316389.1 | - | 10, 11 | 1515 | |||
BC004352.1 | - | 9, 10 | 1479 | 3535435 | ||
BC028155.1 | - | 9, 10 | 1465 | 5241557 | ||
BT007259.1 | - | 9, 10 | 1460 |