Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016876.3 | NP_058572.2 | 6, 7 | 430 | ||
GenBank mRNA | AF108214.1 | - | 6, 7 | 444 | ||
AK038249.1 | - | 6, 7 | 440 | |||
AK168362.1 | - | 6, 7 | 433 | |||
BC008511.1 | - | 6, 7 | 430 | 3592537 | ||
U70733.1 | - | 6, 7 | 420 |