Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016859.3 | NP_058555.3 | 7 | 1307 | ||
GenBank mRNA | AF007802.1 | - | 5 | 536 | ||
AK019239.1 | - | 7 | 1042 | |||
AK032993.1 | - | 7 | 1039 | |||
AK049195.1 | - | 7 | 1300 | |||
AK051704.1 | - | 7 | 1297 | |||
AK083033.1 | - | 7 | 1294 | |||
AK143027.1 | - | 7 | 1308 | |||
AK168301.1 | - | 7 | 1045 | |||
BC017530.1 | - | 7 | 1034 | 4925307 |