Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320611.1 | NP_001307540.1 | 3 | 717 | ||
NM_002266.3 | NP_002257.1 | 3 | 359 | |||
GenBank mRNA | AK024833.1 | - | 3 | 227 | ||
AK058111.1 | - | 3 | 228 | |||
AK291954.1 | - | 3 | 226 | |||
BC005978.1 | - | 3 | 226 | 4105058 | ||
BC013257.1 | - | 3 | 222 | |||
BC034443.1 | - | 3 | 185 | |||
BC053343.1 | - | 3 | 215 | 4450470 | ||
BC067848.1 | - | 3 | 458 | 4825235 | ||
BC146905.1 | - | 3 | 213 | 9056833 | ||
BT006665.1 | - | 2 | 97 | |||
DC388846.1 | - | 3 | 359 | |||
U09559.1 | - | 3 | 229 | |||
U28386.1 | - | 3 | 229 |