Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016897.3 | NP_058593.2 | 6 | 636 | ||
GenBank mRNA | AB021122.1 | - | 6 | 616 | ||
AK013976.1 | - | 6 | 582 | |||
AK146516.1 | - | 6 | 602 | |||
BC037022.1 | - | 7 | 613 | 5366968 | ||
BC062963.1 | - | 6 | 572 | 5692352 | ||
BC069903.1 | - | 6 | 606 | 5290493 | ||
BC096669.1 | - | 6 | 605 | 5056242 | ||
BC132019.1 | - | 6 | 585 | 40130296 | ||
BC132319.1 | - | 6 | 585 | 40130596 |