Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185153.1 | NP_001172082.1 | 2, 3 | 271 | ||
NM_015785.2 | NP_056600.1 | 2, 3 | 442 | |||
GenBank mRNA | BC139450.1 | - | 2, 3 | 411 | 8862472 | |
BC139453.1 | - | 2, 3 | 411 | 8862475 | ||
BC145628.1 | - | 2, 3 | 240 | 9054196 | ||
D17569.1 | - | 2, 3 | 431 |