Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001046637.1 | NP_001040102.1 | 1 | 293 | ||
NM_001285825.1 | NP_001272754.1 | 2 | 367 | |||
NM_015782.3 | NP_056597.3 | 2 | 370 | |||
GenBank mRNA | AK013886.1 | - | 2 | 344 | ||
AK049541.1 | - | 1 | 687 | |||
AK088550.1 | - | 2 | 228 | |||
BC003229.1 | - | 2 | 270 | 3483391 | ||
BC094006.1 | - | 2 | 230 | 4983854 | ||
BC096648.1 | - | 2 | 224 | 1397390 | ||
CA552113.1 | - | 2 | 256 | |||
CX206945.1 | - | 2 | 370 | |||
CX565356.1 | - | 1 | 263 | |||
L15447.1 | - | 2 | 166 |