Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252518.1 | NP_001239447.1 | 4 | 491 | ||
NM_023144.2 | NP_075633.2 | 4 | 494 | |||
GenBank mRNA | AK004830.1 | - | 4 | 457 | ||
AK013444.1 | - | 4 | 408 | |||
AK028338.1 | - | 4 | 459 | |||
AK033201.1 | - | 4 | 420 | |||
AK145179.1 | - | 4 | 467 | |||
AK161152.1 | - | 4 | 478 | |||
AK165855.1 | - | 4 | 453 | |||
AK167297.1 | - | 4 | 425 | |||
AK169202.1 | - | 4 | 457 | |||
AK169303.1 | - | 4 | 478 | |||
AK213551.1 | - | 2 | 317 | |||
BC005465.1 | - | 4 | 453 | 3600935 | ||
BC083074.1 | - | 2 | 331 | 6390386 | ||
CT010241.1 | - | 2 | 313 | |||
S64860.1 | - | 4 | 441 |