Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vti1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016800.3 | NP_058080.2 | 4 | 520 | ||
GenBank mRNA | AF035208.1 | - | 4 | 407 | ||
AF035821.1 | - | 4 | 393 | |||
AK002289.1 | - | 4 | 512 | |||
AK160624.1 | - | 3 | 454 | |||
BC002290.1 | - | 3 | 445 | |||
BC009810.1 | - | 4 | 450 | 4220215 | ||
BC080204.1 | - | 4 | 496 | 3672445 | ||
CX239415.1 | - | 4 | 497 |