Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002268.4 | NP_002259.1 | 15 | 1610 | ||
GenBank mRNA | AB002533.2 | - | 15 | 1313 | ||
AK291041.1 | - | 15 | 1562 | |||
BC028691.2 | - | 15 | 1555 | 4838709 | ||
BC034493.2 | - | 15 | 1610 | 4821602 | ||
U93240.1 | - | 15 | 1454 | |||
Y12393.1 | - | 6 | 584 |