Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169114.1 | NP_001162585.1 | 2 | 245 | ||
NM_016888.5 | NP_058584.3 | 2 | 368 | |||
GenBank mRNA | AF092050.2 | - | 2 | 146 | ||
AK160088.1 | - | 2 | 337 | |||
AY043479.1 | - | 2 | 326 | |||
BC009075.1 | - | 2 | 245 | 2654354 |