Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016928.2 | NP_058624.2 | 4 | 501 | ||
GenBank mRNA | AF186107.1 | - | 1 | 1170 | ||
AK036842.1 | - | 4 | 503 | |||
BC125240.1 | - | 2 | 499 | 40043826 | ||
BC125247.1 | - | 2 | 499 | 40055024 | ||
BC125261.1 | - | 2 | 499 | 40048274 |