Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002269.2 | NP_002260.2 | 6 | 589 | ||
XM_011535805.1 | XP_011534107.1 | 7 | 767 | |||
XM_017010838.1 | XP_016866327.1 | 7 | 767 | |||
XM_017010839.1 | XP_016866328.1 | 6 | 570 | |||
XM_017010840.1 | XP_016866329.1 | 7 | 727 | |||
XM_017010841.1 | XP_016866330.1 | 2 | 69 | |||
XM_017010842.1 | XP_016866331.1 | 6 | 570 | |||
XM_017010843.1 | XP_016866332.1 | 7 | 694 | |||
GenBank mRNA | AF005361.1 | - | 5 | 454 | ||
AK314206.1 | - | 6 | 582 | |||
BC047409.1 | - | 6 | 581 | 5297277 |