Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017390.4 | NP_059086.3 | 2 | 388 | ||
GenBank mRNA | AK035204.1 | - | 2 | 313 | ||
AK040724.1 | - | 2 | 314 | |||
AK090151.1 | - | 2 | 310 | |||
AK090302.1 | - | 2 | 314 | |||
AK136679.1 | - | 2 | 312 | |||
AK144285.1 | - | 2 | 313 | |||
AK144389.1 | - | 2 | 313 | |||
AK144429.1 | - | 2 | 313 | |||
AK144524.1 | - | 2 | 310 | |||
AK165704.1 | - | 2 | 313 | |||
AK165713.1 | - | 2 | 313 | |||
AK165727.1 | - | 2 | 310 | |||
AK165753.1 | - | 2 | 313 | |||
AK165825.1 | - | 2 | 313 | |||
BC107275.1 | - | 2 | 313 | 40052650 | ||
BC107276.1 | - | 2 | 313 | |||
DV056729.1 | - | 2 | 422 |