Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002271.4 | NP_002262.3 | 27 | 3157 | ||
XM_005254052.3 | XP_005254109.1 | 26 | 3067 | |||
XM_005254053.3 | XP_005254110.1 | 26 | 3084 | |||
XM_011521087.2 | XP_011519389.1 | 28 | 3248 | |||
XM_011521088.2 | XP_011519390.1 | 28 | 3314 | |||
XM_011521089.2 | XP_011519391.1 | 29 | 3371 | |||
XM_011521090.2 | XP_011519392.1 | 27 | 3207 | |||
XM_017020562.1 | XP_016876051.1 | 27 | 3188 | |||
GenBank mRNA | AK125031.1 | - | 14 | 2636 | ||
AK225286.1 | - | 24 | 2942 | |||
AK302812.1 | - | 24 | 2929 | |||
AK303494.1 | - | 22 | 2570 | |||
BC000947.2 | - | 5 | 563 | |||
BC001497.2 | - | 24 | 2925 | 2823581 | ||
BC019309.1 | - | 24 | 2925 | |||
BC045640.1 | - | 27 | 3138 | 5298410 | ||
BX648222.1 | - | 24 | 2987 | |||
U72761.1 | - | 24 | 2923 | |||
Y08890.1 | - | 27 | 3159 |