Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004985.4 | NP_004976.2 | 2 | 290 | ||
NM_033360.3 | NP_203524.1 | 2 | 290 | |||
XM_006719069.3 | XP_006719132.1 | 2 | 296 | |||
XM_011520653.2 | XP_011518955.1 | 2 | 282 | |||
XM_017019292.1 | XP_016874781.1 | 2 | 834 | |||
XM_017019293.1 | XP_016874782.1 | 2 | 785 | |||
GenBank mRNA | AF493917.1 | - | 1 | 98 | ||
AK292510.1 | - | 2 | 251 | |||
BC013572.2 | - | 2 | 245 | 3878884 | ||
BT007153.1 | - | 1 | 98 | |||
M35504.1 | - | 1 | 98 | |||
M35505.1 | - | 1 | 98 | |||
M54968.1 | - | 2 | 290 |