Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181649.6 | NP_857632.4 | 4, 5 | 593 | ||
GenBank mRNA | AK009084.1 | - | 4, 5 | 480 | ||
AK148331.1 | - | 4, 5 | 595 | |||
BC047998.1 | - | 4, 5 | 507 | 6511097 | ||
BC092522.1 | - | 4, 5 | 520 | 30648550 | ||
BC138310.1 | - | 4, 5 | 488 | 8861330 | ||
BY741452.1 | - | 4, 5 | 594 |