Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007818.3 | NP_031844.1 | 11 | 1223 | ||
GenBank mRNA | AK004861.2 | - | 2173 | |||
AK149433.1 | - | 11 | 1234 | |||
BC010528.1 | - | 11 | 1223 | 4194909 | ||
BC094062.1 | - | 11 | 1219 | 4162188 | ||
X60452.1 | - | 11 | 1219 |
Gene Symbol: Cyp3a41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017396.3 | NP_059092.2 | 11 | 1232 | ||
GenBank mRNA | AB033414.1 | - | 11 | 1230 |
Gene Symbol: Cyp3a41b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105159.1 | NP_001098629.1 | 11 | 1236 | ||
GenBank mRNA | BC138083.1 | - | 1165 | 8861102 | ||
BC138084.1 | - | 1163 | 8861103 |
Gene Symbol: Cyp3a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177380.3 | NP_796354.1 | 11 | 1230 | ||
GenBank mRNA | AB039380.1 | - | 11 | 1230 | ||
BC125522.1 | - | 11 | 1192 | 40130037 |