Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004985.4 | NP_004976.2 | 4 | 529 | ||
NM_033360.3 | NP_203524.1 | 4 | 529 | |||
XM_006719069.3 | XP_006719132.1 | 4 | 535 | |||
XM_011520653.2 | XP_011518955.1 | 4 | 521 | |||
XM_017019293.1 | XP_016874782.1 | 4 | 1024 | |||
GenBank mRNA | AF493917.1 | - | 3 | 337 | ||
AK292510.1 | - | 4 | 490 | |||
BC013572.2 | - | 4 | 484 | 3878884 | ||
BT007153.1 | - | 3 | 337 | |||
M35504.1 | - | 3 | 337 | |||
M35505.1 | - | 3 | 337 | |||
M54968.1 | - | 4 | 529 |