Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172493.2 | NP_766081.1 | 12 | 1515 | ||
GenBank mRNA | AB713187.1 | - | 13 | 1310 | ||
AK083254.1 | - | 5 | 447 | |||
AK088648.1 | - | 12 | 1449 | |||
AK153986.1 | - | 12 | 1503 | |||
AY312280.1 | - | 12 | 1452 |