Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282072.1 | NP_001269001.1 | 5, 6 | 970 | ||
NM_001282073.1 | NP_001269002.1 | 6, 7 | 894 | |||
NM_001282076.1 | NP_001269005.1 | 6, 7 | 927 | |||
NM_016874.3 | NP_058570.1 | 5, 6 | 970 | |||
GenBank mRNA | AF102818.1 | - | 5, 6 | 830 | ||
AI430175.1 | - | 4, 5 | 452 | |||
AK082115.1 | - | 5, 6 | 823 | |||
AK144333.1 | - | 5, 6 | 739 | |||
AK146546.1 | - | 5, 6 | 847 | |||
AK155836.1 | - | 5, 6 | 936 | |||
AK158586.1 | - | 6, 7 | 779 | |||
BC046399.1 | - | 6, 7 | 872 | 5063995 | ||
FJ377318.1 | - | 6, 7 | 787 | |||
FJ377319.1 | - | 5, 6 | 815 |